You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Safety Clothing
▸
Lab Coats and Jackets
▸
General Purpose Lab Coats and Jackets
▸
Anti-Static Disposable White SMS Lab Coats
Fisher Scientific™
Anti-Static Disposable White SMS Lab Coats
Single use SMS lab coats prevent static discharge, static jump, and static decay
Have Questions?
Change view
Catalog Number
Size
Length (Metric)
Size (Metric) Chest
17100929
X-Large
105 cm
70.5cm
17100928
Large
100 cm
66.5cm
17100931
also known as FIS-17100931
3X-Large
115 cm
78cm
17100930
2X-Large
110 cm
75cm
4 Options
Catalog number
17100929
Price (KRW)
-
Size:
X-Large
Length (Metric):
105 cm
Size (Metric) Chest:
70.5cm
Single use SMS lab coats prevent static discharge, static jump, and static decay
40gsm SMS anti-static material
Knit cuffs and collar
Three pockets
Single Use
Snap closure
White
Specifications
Collar Type
Knit
Latex Free
Yes
Length (Metric)
105 cm
Length (Metric) Sleeve
53cm
Size (Metric) Chest
70.5cm
Closure Type
Snaps
Color
White
Cuff Style
Knit
Disposable
Single-use
Material
SMS
No. of Pockets
3
Product Type
Anti-static Lab Coat
Size
X-Large
Unit Size
Pack of 10