Specifications
Certifications/ComplianceFM, NFPA, OSHA
Depth (English) Interior14.56 in.
Depth (Metric) Exterior45.7 cm
Depth (Metric) Interior37 cm
Dimensions (D x W x H) Interior14.56 x 39.5 x 39.62 in.
Door StyleManual-close
For Use With (Application)Flammables
Height (English) Interior39.62 in.
Height (Metric) Exterior112 cm
Height (Metric) Interior37 cm
IncludesDouble wall, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self latching doors and leveling feet, Polyethylene trays, separate Polyethylene liner
Load Bearing Capacity30 gal.
No. of Doors2
No. of Shelves1
Surface CoatingEpoxy
Weight (English)222 lb.
Weight (Metric)101 kg
Width (English) Interior39.5 in.
Width (Metric) Exterior109 cm
Width (Metric) Interior100.3 cm
ColorBlue
Depth (English) Exterior18 in.
Height (English) Exterior44 in.
MaterialSteel (CR)
Product TypeCorrosives and Acids Storage Cabinet
Width (English) Exterior43 in.
Unit SizeEach