Short-Term Detector Tubes: Water Vapor
Fisher Scientific™

Short-Term Detector Tubes: Water Vapor

Detector tubes
Have Questions?
Catalog NumberMeasurement RangeTube TypeResponse Time
17985321D
also known as FISHERMIS-17985321D-10/PK
5 to 200Water vapor 5/a-p25 min.
Catalog number 17985321D
also known as FISHERMIS-17985321D-10/PK
Price (KRW)
-
Measurement Range:
5 to 200
Tube Type:
Water vapor 5/a-p
Response Time:
25 min.
Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Specifications
Humidity RangeNot Applicable
Measurement Range5 to 200
Quantity10
Response Time25 min.
Temperature Range (English)32°F to 104°F
Temperature Range (Metric)0°C to 40°C
Tube TypeWater vapor 5/a-p
Components MeasuredWater Vapor
Unit SizePack of 10