N95 Dura-Mesh™ Molded Particulate Respirators
Fisher Scientific™
N95 Dura-Mesh™ Molded Particulate Respirators
N95 respirators with latex two-strap design are constructed of a high-resistant shell that holds its shape longer even in hot and humid environments.
Have Questions?
Catalog NumberColorSizeWith Exhalation ValveNuisance Odor Relief
19003246F
also known as FISHERMIS-19003246F-10/PK
GrayMedium/LargeNoOzone and Organic Vapors
Catalog number 19003246F
also known as FISHERMIS-19003246F-10/PK
Price (KRW)
-
Color:
Gray
Size:
Medium/Large
With Exhalation Valve:
No
Nuisance Odor Relief:
Ozone and Organic Vapors
N95 respirators with latex two-strap design are constructed of a high-resistant shell that holds its shape longer even in hot and humid environments.
  • Dura-Mesh™ shell resists collapsing in heat and humidity
  • Molded nose bridge seals easily without a metal noseband
  • Facepiece is naturally contoured for less pressure
  • Straps are securely attached so they don’t break at the seal
  • Softspun™ lining for increased comfort and durability
  • Soft foam nose cushion for added comfort
  • Available in three sizes to fit more workers and increase OSHA compliance
  • NIOSH certified to have a filter efficiency of 95% or greater for non-oil based particulates
  • Certified under 42CFR84
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2009 Section 7.11.1
  • N95 Series Applications: Grinding, torch cutting, sanding, metals, sweeping, bagging, foundries, stone quarrying, agriculture, construction, cement, ceramics manufacturing, underground mining, timber/lumber mills, polishing, buffing, lime, poultry, textiles, welding, brazing, soldering
  • N95 Plus Nuisance Levels of Ozone and Organic Vapors Applications (Fisher cat #19-003-246F only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal
  • N95 Plus Nuisance Levels of Acid Gas Irritants Applications (Fisher cat #19-003-246G only): Aluminum smelting, glass etching, electroplating, fiberglass manufacturing, paper processing, chemical processing, brewing operations, fertilizer production
Specifications
ANSI StandardANSI/ISEA 110-2009, Section 7.11.1
Aerosol TypeN - Not Resistant to Oil
Certifications (NIOSH)NIOSH 42 CFR 84, N95
FDA Cleared Surgical MaskNo
Filter Efficiency95%
For Use With (Application)Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, lime, poultry and textiles
Latex FreeNo
Material HeadstrapLatex
Model2400N95
Nuisance Odor ReliefOzone and Organic Vapors
Quantity10/Pk.
Strap Attachment TypeTwo-Strap Design
ColorGray
Design TypeCup/Molded
DisposableSingle-use
Filter TypeN95
MaterialDura-Mesh
Product TypeParticulate Respirator with Nuisance Organic Vapor Relief
SizeMedium/Large
With Exhalation ValveNo
Unit SizePack of 10