You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Floor Mats
▸
Anti-fatigue Floor Mats
▸
UltraSoft Diamond-Plate Mat
Fisher Scientific™
UltraSoft Diamond-Plate Mat
Antifatigue mat designed for ultimate comfort and performance
Have Questions?
Change view
Catalog Number
Dimensions (L x W)
Color
19042225
also known as FISHERTOPSELLING-19042225-EACH
5 x 3 ft.
Black/Yellow
19042228
also known as FISHERMIS-19042228-EACH
3 x 2 ft.
Black/Yellow
2 Options
Catalog number
19042225
also known as FISHERTOPSELLING-19042225-EACH
Price (KRW)
-
Dimensions (L x W):
5 x 3 ft.
Color:
Black/Yellow
Antifatigue mat designed for ultimate comfort and performance
For heavy use areas, mats resist abrasion and most chemical spills
Double thick sponge base made from 0.94 in. (2.4cm) immediate recovery Nitricell™ sponge
Specially formulated blend of nitrile rubber and PVC
Slip resistant, diamond embossed vinyl surface resists chemicals, abrasion and fire
All edges are beveled for safety and convenience
Recycled: Item contains recycled content as described in the item detail
Specifications
Dimensions (L x W)
5 x 3 ft.
Color
Black/Yellow
Length (Metric)
152.4 cm
Material
Double-thick Nitricell Sponge
Width (Metric)
91.4 cm
Unit Size
Each