You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Atmosphere-Supplying Respirators
▸
Respiratory Air Sources
▸
Respiratory System Air Compressors and Air Pumps
▸
Ambient Air Pump, Model A-750
Ambient Air Pump, Model A-750
Portable and lightweight air source for one hood use, or up to two full- or half-face respirator users
Have Questions?
Catalog Number
Includes
19087451
also known as FISHERMIS-19087451-EACH
Catalog number
19087451
also known as FISHERMIS-19087451-EACH
Price (KRW)
-
Portable and lightweight air source for one hood use, or up to two full- or half-face respirator users
Hansen and Schrader fittings available
1425rpm at 50Hz, 1725 at 60Hz
Working pressure: 2 to 12psi, maximum 15psi
Brass and galvanized steel plumbing
Volts: 15 to 230V, AC single phase
Adjustable brass pressure relief valve
Rigid metal bass and housing
Weight: 24kg (53 lb.)
Powder coated blue
Steel handle
Oil-less
Specifications
Model
9821H
Weight (English)
53 lb.
Applications
Painting or refinishing booths
Material
Metal housing, brass and steel plumbing
Product Type
Low Pressure Air Pump
Unit Size
Each