You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Safety Clothing
▸
Lab Coats and Jackets
▸
General Purpose Lab Coats and Jackets
▸
Men's Poly/Cotton Lab Coats
Fisher Scientific™
Men's Poly/Cotton Lab Coats
With Front gripper closures
Have Questions?
Change view
Catalog Number
Color
Size
Size (English) Chest
19166268
also known as FIS-19166268
White
X-Small
32 to 34 in.
19166260
also known as FIS-19166260
White
Small
36 to 38 in.
2 Options
Catalog number
19166268
also known as FIS-19166268
Price (KRW)
-
Color:
White
Size:
X-Small
Size (English) Chest:
32 to 34 in.
With Front gripper closures
Made of 80/20 polycotton, 5 oz. performance-blend fabric
One-piece, lined, notched lapel collar
Front gripper closures
Two outside lower pockets with one interior left breast pocket
White
Specifications
Collar Type
Notched lapel
Finish
Pre-cure durable press
Length (English)
41.5 in.
Length (Metric)
105.4 cm
Size (English) Chest
32 to 34 in.
Closure Type
Snaps
Color
White
Gender
Men's
Material
Cotton, Polyester
No. of Pockets
3
Size
X-Small
Style
Regular
Unit Size
Each