Specifications
Certifications/ComplianceCE
Fill Volume1 mL
For Use With (Application)Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
IncludesHigh performance large tip purFlock ultra swab, wrapped and 1mL Universal Transport Medium Filled Vial
PackagingIndividually-wrapped
Quantity300 Cs.
Shaft MaterialPolystyrene
Shelf Life18 Months
Storage Requirements2°C to 25°C
Tip TypeLarge Flocked
Volume (Metric)1 mL
Media TypeHank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt
Product TypeUniTranz-RT Transport System
SterilitySterile
Swab ConfigurationSingle Applicator
pH7.3 ±0.2
Unit SizeCase of 300