MiniCollect™ Capillary Blood Collection System Tubes
Fisher Scientific™

MiniCollect™ Capillary Blood Collection System Tubes

Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)AdditiveClosure Color
22030406
also known as FISHERMIS-22030406-100/PK
0.8 mLLithium HeparinGreen
22030402
also known as FISHERMIS-22030402-100/PK
1 mLK3 EDTALavender
Catalog number 22030406
also known as FISHERMIS-22030406-100/PK
Price (KRW)
-
Capacity (Metric):
0.8 mL
Additive:
Lithium Heparin
Closure Color:
Green
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
  •  Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
  • Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
  • Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
  • All tubes require MiniCollect funnel or capillary collection device (sold separately)

 

Specifications
Closure ColorGreen
Closure IncludedYes
For Use With (Application)Small volume blood collection
AdditiveLithium Heparin
Capacity (Metric)0.8 mL
Closure TypeValved Cap
Product TypeMiniCollect Capillary Blood Collection Tube
Unit SizePack of 100