LeLoop™ Inoculating Loops
Fisher Scientific™

LeLoop™ Inoculating Loops

More economical than platinum or disposable plastic loops
Have Questions?
Catalog NumberVolumeCalibratedDiameter (Metric) Inside
22032092
also known as FISHERMIS-22032092-25/PK
3.41 μLNo0.29 cm
Catalog number 22032092
also known as FISHERMIS-22032092-25/PK
Price (KRW)
-
Volume:
3.41 μL
Calibrated:
No
Diameter (Metric) Inside:
0.29 cm
More economical than platinum or disposable plastic loops

Reusable loops are made of high-quality twisted nichrome wire.

  • Loops slide smoothly across agar surfaces without tearing or gouging
  • The nichrome wire cools rapidly, saving time
  • Loops are available calibrated or noncalibrated
  • Calibrated in accordance with ASM/APHA via Evans Blue Test and include calibration certificates
  • LeLoop™ loops come without holders, which are available separately
Specifications
CalibratedNo
DisposableSingle-use
Diameter (English) Inside0.11 in.
Diameter (Metric) Inside0.29 cm
Length (English)3 in.
Length (Metric)7.62 cm
MaterialNichrome
Product TypeInoculating Loop
SizeMedium
SterilityNon-sterile
Volume3.41 μL
Unit SizePack of 25