Sharps-A-Gator™ Sharps Container for Phlebotomy
Sharps-A-Gator™ Sharps Container for Phlebotomy
Sharps-A-Gator™ Sharps Container for Phlebotomy
Fisher Scientific™

Sharps-A-Gator™ Sharps Container for Phlebotomy

Sharps containers for efficient disposal of blood, hypodermic and IV needles
Have Questions?
Catalog NumberCapacity (English)
22037970
also known as FISHERMIS-22037970-EACH
1 qt.
Catalog number 22037970
also known as FISHERMIS-22037970-EACH
Price (KRW)
-
Capacity (English):
1 qt.
For safe, efficient disposal of blood, hypodermic and IV needles.

  • Bright red, one-piece polypropylene container clearly marked with biohazard label
  • Translucent top has convenient holder for blood tubes with a needle remover connected to the holder
  • Sliding lid can snap into open position, slide fully shut without locking, or permanently lock shut
  • Safe to autoclave or incinerate

Specifications
Capacity (Metric)0.9 L
Closure ColorTranslucent
Closure TypeLockable
Dimensions (L x W x H)4.25 x 4.25 x 6.25 in.
DisposableSingle-use
For Use With (Application)Facilitates the disconnection of multi-sample needles from needle holders
Height (English)6.25 in.
Height (Metric)15.9 cm
Length (English)4.25 in.
Length (Metric)10.8 cm
Quantity18/Cs.
Width (English)4.25 in.
Width (Metric)10.8 cm
AutoclavableAutoclavable
Capacity (English)1 qt.
ColorRed
MaterialPolypropylene
Unit SizeEach