Chlorine TRACER PockeTester
Fisher Scientific™

Chlorine TRACER PockeTester

TRACER PockeTester Total Chlorine Kit, Read Total Chlorine from 0.00 to 9.99ppm to 0.01ppm resolution, Extra bold display includes an analog bar graph feature, uses four LR-44 batteries, Readings are not affected by sample color
Have Questions?
Catalog NumberData Points
22278617
also known as FISHERMIS-22278617-EACH
15
Catalog number 22278617
also known as FISHERMIS-22278617-EACH
Price (KRW)
-
Data Points:
15
TRACER PockeTester Total Chlorine Kit, Read Total Chlorine from 0.00 to 9.99ppm to 0.01ppm resolution, Extra bold display includes an analog bar graph feature, uses four LR-44 batteries, Readings are not affected by sample color

Reads Total Chlorine from 0.00 to 9.99ppm to 0.01ppm resolution, Extra bold display includes an analog bar graph feature, uses four LR-44 batteries. Readings are not affected by sample color.

  • Large 0.75L x 0.93 in.W (1.9 x 2.8cm) LCD display with analog bar graph
  • Memory saves calibration values and stores up to 15 readings
  • Sample color or turbidity do not affect readings
  • Sample temperature can be displayed in chlorine mode, or in pH mode when using optional pH sensor probe
  • Automatically self-calibrates and shuts off when not in use; also indicates when batteries are low

Specifications
Data Points15
Display TypeLCD
For Use With (Application)Industrial water boiling and cooling, laundry and sanitation, water and wastewater
Includes100 reagent tablets that deliver a precise amount of iodide for a 20mL sample, and four LR-44 batteries. Optional accessories include pH, ORP, and Cl2; sensor probes, and weighted stand with 5 sample cups.
Length (Metric)18.4 cm
Resolution0 to 9.99 to 0.01 ppm
Temperature (English) Operating23°F to 194°F
Width (English)0.93 in.
Width (Metric)2.8 cm
Detection Range0.00 to 9.99 ppm
TypeChlorine Tracer Pocket Tester
Unit SizeEach