Specifications
For Use With (Application)Easy-to-open tear line
Height (English)6 in.
Height (Metric)15.24 cm
IncludesBlack and orange biohazard warning, double pocket, temperature indicators and tear line
Width (Metric)10.16 cm
ColorClear
Product TypeReclosable Biohazard Bag
Width (English)4 in.
Unit SizeCase of 1000