You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
First Aid and Medical
▸
Patient Care Products
▸
Cotton Balls and Packing
▸
Nonsterile Cotton Balls
Fisher Scientific™
Nonsterile Cotton Balls
Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.
Have Questions?
Change view
Catalog Number
Weight (Metric)
Quantity
Size
22456880
also known as FIS-22456880
0.45 kg
4000 Cs.
Medium
22456882
1.08 kg
2000 Cs.
X-Large
2 Options
Catalog number
22456880
also known as FIS-22456880
Price (KRW)
-
Weight (Metric):
0.45 kg
Quantity:
4000 Cs.
Size:
Medium
Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.
Uniform in size and shape
Made of pure, natural, bleached cotton for increased absorbency
Available in sizes medium, large, extra large and bulk roll
Specifications
Quantity
4000 Cs.
Size
Medium
Sterility
Non-sterile
Weight (English)
0.006 lb.
Weight (Metric)
0.45 kg
Product Type
Cotton Balls
Unit Size
Case of 4000