T-Shaped Spreaders
Fisher Scientific™
T-Shaped Spreaders
T-shaped spreaders
Have Questions?
Catalog NumberQuantity
23600896
also known as FIS-23600896
10/Peel Pk., 500 Spreaders/Box
Catalog number 23600896
also known as FIS-23600896
Price (KRW)
-
Quantity:
10/Peel Pk., 500 Spreaders/Box
T-shaped spreaders
T-Shaped Plastic Spreaders
  • Designed for spreading and dispersing liquids across the surface of agar culture plates
  • Used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples
  • Smooth, rounded surfaces prevent any cutting or gouging of agar during inoculation of culture plates
  • T-Shape design means that even pressure is applied across the entire length of the T-bar in contact with the agar
  • Available individually sterile wrapped or in packs of five or ten pieces in a peel pack
Specifications
For Use With (Application)Designed for spreading and dispersing liquids across the surface of agar culture plates, used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples
For Use With (Equipment)Culture Plates
Quantity10/Peel Pk., 500 Spreaders/Box
ShapeT-Shape
MaterialPlastic
Product TypeCell Spreader
SterilitySterile
Unit SizeCase of 500