You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Histology and Cytology
▸
Microtomes and Accessories
▸
Ultra Disposable Microtome Blades
Ultra Disposable Microtome Blades
Epredia™ ultra microtome blades offer high consistency, impressive durability, dependability, and precise performance.
Have Questions?
Catalog Number
Profile
3053835
Low
Catalog number
3053835
Price (KRW)
-
Profile:
Low
High- and low-profile universal blades can be used with any microtome or cryostat on the market
Unique angle profile that facilitates faster ribboning
Increased Longevity: Produces an edge that is twice as strong as traditional blades
Reliable Consistency: New proprietary process ensures a durable and consistent coating
Specifications
Cutting Angle
34°
Disposable
Single-use
For Use With (Equipment)
Microtomes, Cryostats
Profile
Low
Unit Size
Case of 50