Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg
Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg
Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg
Fisher Scientific™

Potassium Phosphate, Monobasic, Molecular Biology Grade, Calbiochem™, , 1kg

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 5295681KG
also known as FISHERMIS-5295681KG-EACH
Price (KRW)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7778-77-0
Specifications
Assay Percent Range≥99%
CAS7778-77-0
ChEBICHEBI:63036
Chemical Name or MaterialPotassium Phosphate, Monobasic
EINECS Number231-913-4
View more