Search
| Catalog Number | Plug Type | Includes |
|---|---|---|
| 700611110 also known as LABCONCO-700611110-EACH | North America | Stainless Steel Collector, Purge Valve |
| 700612110 also known as LABCONCO-700612110-EACH | North America | PTFE-coated Collector, Purge Valve |
FreeZone 6L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for a vacuum pump and accommodates the FreeZone Stoppering Tray Dryer (pump and tray dryer sold separately).
These models offer the broadest range of built-in options including a shell freezer.
Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).