Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Fisher Scientific™
Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
AS2752.8cm wide plastic shell vials
AS273
also known as FISHERTOPSELLING-AS273-EACH
2.5cm narrow plastic shell vials
AS271Standard glass vials
AS277Standard plastic stock bottles
Catalog number AS275
Price (KRW)
-
For Use With (Equipment):
2.8cm wide plastic shell vials
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
  • Superior barrier to mite infestation
  • Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
  • Stretches yet maintains tight fit after repeated removal and closing
  • Flat surface accepts marking pen
Specifications
For Use With (Application)Minimizes evaporation, keeps food fresh longer
For Use With (Equipment)2.8cm wide plastic shell vials
IncludesSuperior barrier
Quantity1000 Case
MaterialCellulose Acetate
TypeBuzzPlugs Bottle Closure
Unit SizeCase of 1000