Drosophila Vials
Fisher Scientific™
Drosophila Vials
Straight wall design minimizes the dislodging of food; provides more efficient racking, filling and plugging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) OuterTypeMaterialMouth Style
AS51029.21 mmDrosophila Vial, Tray PackedK-ResinWide
AS507
also known as FISHERTOPSELLING-AS507-500/CS
25.12 mmDrosophila Vial, Bulk PackedPolypropyleneNarrow
AS51329.21 mmFly Drosophila VialPolypropyleneWide
AS508
also known as FISHERMIS-AS508-500/CS
25.12 mmDrosophila Vial, Tray PackedK-ResinNarrow
AS51525.12 mmDrosophila VialPolystyreneNarrow
AS51929.21 mmDrosophila Vial, Bulk PackedPolystyreneWide
AS50925.12 mmDrosophila Vial, Bulk PackedK-ResinNarrow
AS51129.21 mmDrosophila Vial, Bulk PackedK-ResinWide
AS51625.12 mmDrosophila Vial, Tray PackedPolystyreneNarrow
AS52029.21 mmDrosophila Vial, Tray PackedPolystyreneWide
AS51229.21 mmDrosophila Vial, Tray PackedPolypropyleneWide
Catalog number AS510
Price (KRW)
319,000
Case of 500
Diameter (Metric) Outer:
29.21 mm
Type:
Drosophila Vial, Tray Packed
Material:
K-Resin
Mouth Style:
Wide
Price (KRW)
319,000
Case of 500
Straight wall design minimizes the dislodging of food; provides more efficient racking, filling and plugging
Fisherbrand Drosophila vials are available in clear polystyrene, translucent and flexible polypropylene, and virtually unbreakable K-resin. These vials feature a flat bottom, straight wall design for convenient racking, filling and stacking. Available bulk packaged or in a convenient, 10 x 10 tray pack configuration.
Polypropylene Vials
  • Translucent and flexible
  • Autoclavable
Polystyrene Vials
  • Excellent clarity for optimal visibility
  • Nonautoclavable
K-Resin Vials
  • Highly flexible, virtually unbreakable
  • Nonautoclavable
Standard-Packed Glass Shell Vials
  • Type 1, N-51A borosilicate glass
  • Securely packed in cardboard trays with individual vial dividers
Bulk- and Tray-Packed Shell Vials
  • Available in narrow- or wide-mouth styles
  • Constructed from polystyrene, K-Resin™, or polypropylene
  • K-Resin and polypropylene vials are virtually unbreakable, scratch and mar resistant; polystyrene and K-Resin vials have glass-like clarity
  • Polypropylene vials are autoclavable; polystyrene and K-Resin are nonautoclavable
Specifications
AutoclavableNot Autoclavable
ColorClear
Diameter (English) Outer1.15 in.
Diameter (Metric) Outer29.21 mm
For Use With (Application)Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
Height (English)3.74 in.
Height (Metric)95 mm
Mouth StyleWide
MaterialK-Resin
TypeDrosophila Vial, Tray Packed
Unit SizeCase of 500