PTFE boiling stones promote efficient boiling, prevent superheating, and minimize bumping during reflux and speed distillation. Stones will not scratch or abrade glassware.
Have Questions?
Catalog Number
Quantity
CP0119900
also known as CP-CP0119900-EACH
450 g
Catalog number CP0119900
also known as CP-CP0119900-EACH
Price (KRW)
-
Quantity:
450 g
PTFE boiling stones promote efficient boiling, prevent superheating, and minimize bumping during reflux and speed distillation. Stones will not scratch or abrade glassware.