Motic Compound Microscope with Digital Camera, Koehler illumination
Cole-Parmer™
Motic Compound Microscope with Digital Camera, Koehler illumination
Color-corrected infinity optical system for high-image quality and flatness of field
Have Questions?
Catalog NumberHead Style
CP0391145
also known as CP-CP0391145-EACH
Binocular
Catalog number CP0391145
also known as CP-CP0391145-EACH
Price (KRW)
-
Head Style:
Binocular
Color-corrected infinity optical system for high-image quality and flatness of field
Color-corrected infinity optical system for high-image quality and flatness of field
  • Stream live images to your PC
Specifications
Data Transfer480 MB/sec
DescriptionCompound Microscope with Digital Camera, Koehler illumination
Dimensions (L x W x H)160 x 140 mm
Frequency50/60 Hz
OutputUSB
Resolution3.0 MP
Voltage100/240 V
Head StyleBinocular
Lighting TypeKoehler Halogen with Rheostat 30w
Magnification Power40X, 100X, 400X, 1000X
Objective TypesAchromatic
Product TypeMicroscope with Digital Camera
Unit SizeEach