Product Image
Cole-Parmer™
316 Stainless Steel Shaft, 3/8 in. dia. x 30 in. L
Accepts 5/16 in. bore diameter propeller
Have Questions?
Catalog NumberType
CP0455205
also known as CP-CP0455205-EACH
Stirring Shaft
Catalog number CP0455205
also known as CP-CP0455205-EACH
Price (KRW)
-
Type:
Stirring Shaft
Accepts 5/16 in.bore diameter propeller
Specifications
Description316 Stainless Steel Extension Rod, 5/16in. dia x 12in. L
MaterialStainless Steel (316)
TypeStirring Shaft
Unit SizeEach