Search
| Catalog Number | Diameter (English) | Length (English) |
|---|---|---|
| CP0477510 also known as CP-CP0477510-EACH | 0.1875 in. | 4375 in. |
| CP0477575 also known as CP-CP0477575-EACH | 0.375 in. | 2 1/2 in. |
Mono-Mold magnetic stir bars are the standard all-purpose stir bar. Available with or without spinning rings; rings reduce contact area between stir bar and vessel, reducing friction and noise.