Cole-Parmer™ pH Probe, Refillable/SJ/Glass/Extra-Long/Thin/326x8mm, BNC
Cole-Parmer™

Cole-Parmer™ pH Probe, Refillable/SJ/Glass/Extra-Long/Thin/326x8mm, BNC

Faster response and increased stability across a pH range of 0 to 14
Have Questions?
Catalog NumberConnector Types
CP0599040
also known as CP-CP0599040-EACH
BNC
Catalog number CP0599040
also known as CP-CP0599040-EACH
Price (KRW)
-
Connector Types:
BNC
Faster response and increased stability across a pH range of 0 to 14

Faster response and increased stability across a pH range of 0 to 14

  • Dome bulb offers better protection from breakage
  • Stability in low-ionic strength solutions
  • Offers resistance to Na+ ion interference
  • Compatible with most brands of pH meters (that accept standard BNC connections)
Specifications
For Use With (Application)Extra-long neck for flasks and bottles
Connector TypesBNC
JunctionSingle
TypepH Probe
pH Range0 to 14
Unit SizeEach