Search
| Catalog Number | Capacity (Metric) |
|---|---|
| CP0610560 also known as CP-CP0610560-4/PK | 100 mL |
Choose from PPCO or PC. Bottles are injection blown—no pinchmarks that could contribute to failure. To use bottles at maximum speeds without leakage or to spin hazardous materials, order sealing caps under accessories. Models 06106-40 and 06105-60 fit the following rotors: Beckman JS-5.2, JS-4.2, JS-4.0, and JS-3.0 (when used with Beckman 344040 adapter). All models fit Sorvall HG-4L, H-4000, H-6000A (no adapter needed) and IEC 266, 949, and 966 (when used with appropriate IEC adapter). Thermo Scientific Nalgene™.