Lazy Susan Design 94-Holder Rotary Pipette Stand
Cole-Parmer™
Lazy Susan Design 94-Holder Rotary Pipette Stand
Pipette stands are constructed of polypropylene.
Have Questions?
Catalog NumberHolds
CP0621720
also known as CP-CP0621720-EACH
Tubes (94)
Catalog number CP0621720
also known as CP-CP0621720-EACH
Price (KRW)
-
Holds:
Tubes (94)
Pipette stands are constructed of polypropylene.
B) Circular Pipette Stands
Pipette stands are constructed of polypropylene. Stand 06217-10 holds 28 pipettes (ten 14-mm dia, eighteen 11-mm dia). Pipettes drain to table. Stand 06217-20 features a lazy Susan design for 94 pipettes. Pipettes drain into troughs on lower disc. Bel-Art.
Specifications
DescriptionRotary Pipette Stand Features A Lazy Susan Design; Holds 94 Pipettes.
Height (English)18.75 in.
Length (English)9 in.
HoldsTubes (94)
MaterialPolypropylene
Product TypePipet Stand
Unit SizeEach