Search
| Catalog Number | Diameter (Metric) Plug |
|---|---|
| CP0629876 also known as CP-CP0629876-200/PK | 27 to 34 mm |
| CP0629878 also known as CP-CP0629878-100/PK | 34 to 45 mm |
Durable and reusable foam stoppers are an economical way to keep the contents of your containers secure. Open-cell foam has a uniform density, leaves no fibers like cotton, is breathable, nonabsorbent, chemcial resistant, heat tolerant, and retains shape after repeated use. Can also be used as a cleaner for pipettes, tubes, vials and precision tubing. Autoclavable.