Masterflex™ Two-Stop Pump Tubing, Pilicone (Platinum-Cured)
Actual product may vary
Cole-Parmer™
Masterflex™ Two-Stop Pump Tubing, Pilicone (Platinum-Cured)
For use with Masterflex™ multichannel pump heads
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeFlow Rate
CP0642114
also known as CP-CP0642114-8/PK
14130mL/min.
CP0642124
also known as CP-CP0642124-4/PK
241800mL/min.
CP0642125
also known as CP-CP0642125-8/PK
251000mL/min.
Catalog number CP0642114
also known as CP-CP0642114-8/PK
Price (KRW)
-
Tubing Size:
14
Flow Rate:
130mL/min.
For use with Masterflex™ multichannel pump heads
For use with Masterflex™ multichannel pump heads
Specifications
Flow Rate130mL/min.
Hose Barb1/16
Tubing Size14
Diameter (English) Outer0.546 in.
Diameter (Metric) Outer14 mm
MaterialSilicone
Product TypePump Tubing
Unit SizePack of 8