Masterflex™ Tygon™ E-Lab (E-3603) Pump Tubing
Actual product may vary
Cole-Parmer™
Masterflex™ Tygon™ E-Lab (E-3603) Pump Tubing
Ideal for general transfer applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeFlow RateHose Barb
CP0650918
also known as CP-CP0650918-EACH
182300mL/min.3/8
CP0650913
also known as CP-CP0650913-EACH
1336mL/min.1/16
CP0650914
also known as CP-CP0650914-EACH
14130mL/min.1/16
CP0650916
also known as CP-CP0650916-EACH
480 mL/min.1/8 in.
CP0650917
also known as CP-CP0650917-EACH
171700mL/min.1/4
CP0650925
also known as CP-CP0650925-EACH
251000mL/min.3/16
Catalog number CP0650918
also known as CP-CP0650918-EACH
Price (KRW)
-
Tubing Size:
18
Flow Rate:
2300mL/min.
Hose Barb:
3/8
Ideal for general transfer applications
Ideal for general transfer applications
  • Inexpensive tubing for general laboratory applications.
  • Clear for easy flow monitoring.
  • Handles virtually all inorganic chemicals.
  • Nonaging, nonoxidizing.
  • Low gas permeability.
Specifications
Flow Rate2300mL/min.
Hose Barb3/8
Tubing Size18
ColorClear
Diameter (English) Outer0.7 in.
Diameter (Metric) Outer18 mm
Product TypePump Tubing
Unit SizeEach