PFA Compression Nut Assembly
PFA Compression Nut Assembly
Actual product may vary
Cole-Parmer™
PFA Compression Nut Assembly
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) Inner
CP3132186
also known as CP-CP3132186-EACH
1/4 in.
CP3132187
also known as CP-CP3132187-EACH
3/8 in.
CP3132188
also known as CP-CP3132188-EACH
1/2 in.
Catalog number CP3132186
also known as CP-CP3132186-EACH
Price (KRW)
-
Diameter (English) Inner:
1/4 in.
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
Specifications
Sterilization MethodAutoclave or Ethylene Oxide (ETO)
Diameter (English) Inner1/4 in.
MaterialPFA
Product TypeCompression Nut Assembly
Unit SizeEach