QRP Incorporated PolyTuff Heat Gloves, 12 in.
Cole-Parmer™
QRP Incorporated PolyTuff Heat Gloves, 12 in.
Ideal for use with diffusion furnaces, burn-in ovens, ceramics, and food processing and handling.
Have Questions?
Catalog number CP3368931
also known as CP-CP3368931-EACH
Price (KRW)
-
Ideal for use with diffusion furnaces, burn-in ovens, ceramics, and food processing and handling.
Specifications
Length (English)12 in.
MaterialPolytuff Silicon
SizeLarge
Unit SizeEach