pHydrion Plastic pH Indicator Strips
Actual product may vary
pHydrion Plastic pH Indicator Strips
pHydrion Plastic pH Indicator Strips
Cole-Parmer™

pHydrion Plastic pH Indicator Strips

Three times the matching area compared with other plastic strips
Have Questions?
Change viewbuttonViewtableView
Catalog NumberpH RangePackaging
CP5920014
also known as CP-CP5920014-100/PK
5 to 9
CP5920010
also known as CP-CP5920010-100/PK
0 to 14Clear Plastic Case
CP5920012
also known as CP-CP5920012-100/PK
0 to 6
CP5920016
also known as CP-CP5920016-100/PK
6.5 to 13
CP5920040
also known as CP-CP5920040-100/PK
0 to 14Flip Top Vial
CP5920048
also known as CP-CP5920048-100/PK
5.5 to 8
Catalog number CP5920014
also known as CP-CP5920014-100/PK
Price (KRW)
-
pH Range:
5 to 9
Three times the matching area compared with other plastic strips

Three times the matching area compared with other plastic strips

Specifications
For Use With (Application)pH testing
FormatStrip
Quantity100 Tests/Kit
TypepH Strip
pH Range5 to 9
Unit SizePack of 100