Masterflex™ Platinum-Cured Silicone Tubing
Actual product may vary
Cole-Parmer™
Masterflex™ Platinum-Cured Silicone Tubing
Nontoxic and nonhemolytic with excellent biocompatibility
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeFlow RateSterilization Method
CP9651089
also known as CP-CP9651089-EACH
8919,000 mL/min.Autoclave, EtO, Gamma Irradiation
CP9651088
also known as CP-CP9651088-EACH
8817,000 mL/min.Autoclave, EtO, Gamma Irradiation
Catalog number CP9651089
also known as CP-CP9651089-EACH
Price (KRW)
-
Tubing Size:
89
Flow Rate:
19,000 mL/min.
Sterilization Method:
Autoclave, EtO, Gamma Irradiation
Nontoxic and nonhemolytic with excellent biocompatibility
  • Nontoxic and nonhemolytic with excellent biocompatibility
  • Long pumping life; pump at continuous pressures up to 40 psi
  • Low extractables and low gas permeability
  • Meets USP Class VI criteria, and is FDA compliant
Specifications
Flow Rate19,000 mL/min.
Hose Barb5/8
Maximum Pressure40 psi
Sterilization MethodAutoclave, EtO, Gamma Irradiation
Tubing Size89
Diameter (English) Outer3.5 in.
Diameter (Metric) Outer89 mm
MaterialSilicone
Product TypePump Tubing
Unit SizeEach