Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Polypropylene Disposable Beakers
Fisher Scientific™
Polypropylene Disposable Beakers
Highly resistant to strong acids, organic solvents and other common laboratory reagents
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FB012918
also known as FIS-FB012918
15 mL
FB012919
also known as FIS-FB012919
30 mL
FB0129110
also known as FIS-FB0129110
50 mL
FB012917
also known as FIS-FB012917
100 mL
FB012915250 mL
FB0129112
also known as FIS-FB0129112
400 mL
FB012916
also known as FIS-FB012916
600 mL
Catalog number FB012918
also known as FIS-FB012918
Price (KRW)
-
Capacity (Metric):
15 mL
Rib construction permits stacking without binding
  • Special “dripless” lip
  • Base allows beaker to sit flat during use
  • Graduated in mL and oz.
Specifications
For Use With (Application)Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents
IncludesSpout
Increments5 mL
TypeBeaker
AutoclavableAutoclavable
Capacity (Metric)15 mL
DisposableSingle-use
GraduatedYes
MaterialPolypropylene
Unit SizePack of 100