5-Place Slide Mailer, End Opening
5-Place Slide Mailer, End Opening
Fisher Scientific™
5-Place Slide Mailer, End Opening
Ship or store slides safely. Fisherbrand™ Five-Slide Mailer is constructed of extra-strong, clear polypropylene with inner channels that keep the slides from touching one another. Snap-top lid closes securely to keep slides safe. Disposable and convenient.
Have Questions?
Catalog number HS15986
also known as FISHERMIS-HS15986-EACH
Price (KRW)
35,000
Each
Add to cart
Price (KRW)
35,000
Each
Add to cart
Ship or store slides safely. Fisherbrand™ Five-Slide Mailer is constructed of extra-strong, clear polypropylene with inner channels that keep the slides from touching one another. Snap-top lid closes securely to keep slides safe. Disposable and convenient.
  • Mailer holds five standard slides (25 x 75 mm and 1 x 3 in)
  • Inside wall features slotted channels to keep slides separated and to make removal easier
  • Lid closes securely to keep slides safe
  • Disposable
Specifications
Closure TypeSnap-top
ColorNatural
Dimensions (L x W x H)43 x 24 x 88 mm
For Use With (Equipment)Standard Slides
MaterialPolypropylene
Quantity25 Pk.
Slide Capacity5
TypeSlide Mailer
Unit SizeEach