Search
| Catalog Number | Length (Metric) |
|---|---|
| K4164300000 also known as FISHERMIS-K4164300000-EACH | 25 cm |
To operate, the streaker is placed over the edge of the counter top and the plate to be streaked is positioned parallel to it. The bent capillary tip is then adjusted so that it lightly touches the plate. The reservoir is filled with the solution to be chromatographed, and by moving the unit back and forth along the counter edge, a uniform streak is accomplished.