Kimble™ Kontes™ Ultra-Ware™ Three-Hole Cap Systems
Fisher Scientific™
Kimble™ Kontes™ Ultra-Ware™ Three-Hole Cap Systems
Borosilicate glass and PTFE construction give excellent chemical resistance to HPLC mobile phases
Have Questions?
Catalog NumberThread Size (Metric)
K9539135000
also known as FISHERMIS-K9539135000-EACH
GL 45
Catalog number K9539135000
also known as FISHERMIS-K9539135000-EACH
Price (KRW)
-
Thread Size (Metric):
GL 45
Borosilicate glass and PTFE construction give excellent chemical resistance to HPLC mobile phases
  • Supplied with three-hole cap, conical bottom reservoir, standard fittings kit, and PTFE tubing
  • Reservoir has autoclavable plastic safety coating that absorbs UV up to 385nm—preserves UV-sensitive mobile-phase components
  • PTFE cap body is supplied with a TFE/propylene O-ring
  • Connecting threads on top and bottom of cap use 1/4-28 flangeless fittings and adapters Filtration of the mobile phase directly into the reservoir requires additional purchase of a filtration cap and appropriate solvent pickup adapter
Specifications
AutoclavableAutoclavable
Neck Finish53 mm
No. per Case1 Cs.
Thread Size (Metric)GL 45
MaterialPTFE
Product TypeMultiple Distributor Screw Cap
Unit SizeEach