Legionella GVPC Agar (ISO) for Water Testing
Legionella GVPC Agar (ISO) for Water Testing
Legionella GVPC Agar (ISO) for Water Testing
Legionella GVPC Agar (ISO) for Water Testing
Thermo Scientific™

Legionella GVPC Agar (ISO) for Water Testing

Isolate Legionellaceae in environmental samples with this selective medium which conforms to ISO standards and yields optimal recovery of Legionellaceae.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberManufacturing Origin
PO1291AUnited Kingdom
PO5374AGermany
Catalog number PO1291A
Price (KRW)
-
Manufacturing Origin:
United Kingdom

Isolate Legionella from water samples using Thermo Scientific™ Legionella GVPC Agar (ISO) for Water Testing, a selective medium that conforms to ISO11731:20171 Water quality – Enumeration of Legionella and shows optimal recovery of Legionellaceae in short incubation periods from environmental samples.Save resources on QC testing with as the medium is membrane filter tested in accordance with ISO 11133:20142 and displays the batch number of the filters used on the Certificate of Analysis. Using the same filter batch with the medium for your water testing removes the need for in-house QC.

Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Legionella is commonly transmitted through aerosols generated from cooling towers of air-conditioning systems or by use of potable water from domestic hot-water installations, for example, showers. Legionella GVPC Agar (ISO) for Water Testing is a recommended  medium for the isolation of Legionella from water samples:

  • Yields optimal recovery in a short incubation period.
  • Conforms to ISO11731:2017 standards.
  • Quality control tested according to ISO11133:2014, including membrane filtration method where the batch number is displayed on the certificate of analysis
  • High sensitivity for recovery of strains such as anisa
  • Highly selective for inhibition of non-target organisms such as Escherichia coli and Enterococcus faecalis
  • Reduces in-house QC requirements if purchased with the associated filter batch.
  • Easy-to-read results as Legionella form grey-blue colonies.
  • Ready-to-use convenience of prepared media. 

Legionella GVPC Agar (ISO) for Water Testing has been formulated for improved detection of sensitive strains like L. anisa, and for inhibition of contaminating flora including E. coli and Enterococcus faecaliswhich if not suppressed can over-grow the target and result in a false-negative result being reported.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

For Laboratory Use
Specifications
ColorTraffic Black to Jet black, opaque
DescriptionLegionella GVPC Agar (ISO) for Water Testing
Fill Volume22 mL
Fill Weight22g
Format90mm Monoplate
Packaging TypeFlow wrapped – single layer
Quantity10/Pk.
Shelf Life12 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach