Remel™ MacConkey Agar
Thermo Scientific™
Remel™ MacConkey Agar
The Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R453802500 g
R45380610 kg
Catalog number R453802
Price (KRW)
-
Quantity:
500 g
Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.
Specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF, SMWW, USP.
DescriptionMacConkey Agar
Media TypeMacConkey Agar
Quantity500 g
FormPowder
Product TypeNutrient Agar
Unit SizeEach