Salmonella Shigella Agar (Dehydrated)
Salmonella Shigella Agar (Dehydrated)
Salmonella Shigella Agar (Dehydrated)
Thermo Scientific™
Salmonella Shigella Agar (Dehydrated)
Perform selective and differential isolation of Salmonella and partly Shigella spp. from clinical specimens using Salmonella Shigella Agar.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R45450610 kg
R454502500 g
Catalog number R454506
Price (KRW)
-
Quantity:
10 kg
Perform selective and differential isolation of Salmonella and partly Shigella spp. from clinical specimens, including feces, and food samples1 using Thermo Scientific™ Remel™ Salmonella Shigella Agar (Dehydrated). Leifson modified the Deoxycholate-Citrate Agar used for the detection of enteric pathogens2 and added bile salts mixture to inhibit the growth of gram-positive organisms.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
Perform selective and differential isolation of Salmonella and partly Shigella spp. from clinical specimens, including feces, and food samples using Salmonella Shigella Agar.
  • Ease of Differentiation: Based on the fermentation of lactose; gram-negative bacilli produce pink to red colonies whereas lactose non-fermenters produce transparent to colorless colonies.
  • Selective: Due to the inclusion of selective inhibitory substances brilliant green, bile salts, thiosulphate, and citrate, most gram-positive and coliform organisms are inhibited.
This medium is used for qualitative procedures. This medium contains bile salts, brilliant green and citrates which acts as selective agents and inhibit gram-positive bacteria and coliforms. Sodium thiosulfate, a sulfur source, and ferric ammonium citrate work as indicators to enable organisms which produce H2S to form black-centered colonies, including some strains of Salmonella. Lactose acts as a differentiator.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
REF R119756 is not intended for IVD use.
  1. Liefson, E., 1935. J. Pathol. Bacteriol. 40:581.
  2. Macfaddin, J.F. 1985. Media for Isolation-Cultivation-Identification-Maintenance of Medical Bacteria. Vol. 1. Williams and Wilkins, Baltimore, MD.
Specifications
Certifications/ComplianceIndustrial Reference: COMPF, USDA.
DescriptionSalmonella Shigella Agar
Quantity10 kg
FormPowder
Product TypeAgar
Unit SizeEach