Product Image
Thermo Scientific™
Remel™ Tryptic Soy Broth
Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R4550542.5 kg
R455052500 g
R45505610 kg
Catalog number R455054
Price (KRW)
-
Quantity:
2.5 kg
Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.
For Laboratory Use Only
Specifications
Certifications/ComplianceIndustrial Reference: USP
DescriptionTryptic Soy Broth (TSB) (Soybean Casein Digest)
Quantity2.5 kg
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach