Product Image
Oxoid UK

OCLA (ISO) Differential Supplement 10 vials

For addition to Chromogenic Listeria Agar (CM1084) for the isolation and enumeration of Listeria sp from food samples
Have Questions?
Catalog number SR0244E
Price (KRW)
-
One vial required for 500 ml of medium
For Laboratory Use
For Laboratory Use
Specifications
Country of OriginUK
For Use With (Application)Supplement for Chromogenic Listeria Agar (CM1084)
FormatBox
Media TypeSupplement
Packaging Type10 vials
Quantity10/Pk.
Shelf Life6 months
Product TypeMicrobiology Media
SpeciesListeria
Unit SizeEach
Contents & Storage
2°C to 8°C