QSP Low Retention Filtered And Non-Filtered Pipette Tips
Thermo Scientific™

QSP Low Retention Filtered And Non-Filtered Pipette Tips

Low retention pipette tips with a hydrophobic inner surface to reduce liquid retention and maintain pipetting precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Tip StyleFilteredUnit Size
TLR070-QPK
also known as TLR070-Q
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 1000
TLR102-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredCase of 10000
TLR102-QPK
also known as TLR102-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredPack of 1000
TLR102R-QPK
also known as TLR102R-Q
QSP 10XL, Non-Filtered10 μLExtended Length, ConicalNon-filteredPack of 960
TLR102RS-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredCase of 4800
TLR102RS-QPK
also known as TLR102RS-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredPack of 960
TLR102RL-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredCase of 4800
TLR102RL-QPK
also known as TLR102RL-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredPack of 960
TLR102RLS-QQSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredCase of 4800
TLR102RLS-QPK
also known as TLR102RLS-Q
QSP 10XL, Non-Filtered10 μLExtended LengthNon-filteredPack of 960
TFLR113-20-Q20 μLBeveledFilteredCase of 4800
TFLR113-20-QPK
also known as TFLR113-20-Q
20 μLBeveledFilteredPack of 960
TFLR113-100-Q100 μLBeveledFilteredCase of 4800
TFLR113-100-QPK
also known as TFLR113-100-Q
100 μLBeveledFilteredPack of 960
TLR070-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 10000
TLR070R-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 4800
TLR070R-QPK
also known as TLR070R-Q
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 960
TLR070RS-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 4800
TLR070RS-QPK
also known as TLR070RS-Q
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 960
TLR070RL-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 4800
TLR070RL-QPK
also known as TLR070RL-Q
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 960
TLR070RLS-QQSP 200, Non-Filtered200 μLBeveledNon-filteredCase of 4800
TLR070RLS-QPK
also known as TLR070RLS-Q
QSP 200, Non-Filtered200 μLBeveledNon-filteredPack of 960
TLR106-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 10000
TLR106-QPK
also known as TLR106-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 1000
TLR106R-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 4800
TLR106R-QPK
also known as TLR106R-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 960
TLR106RS-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 4800
TLR106RS-QPK
also known as TLR106RS-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 960
TLR106RL-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 4800
TLR106RL-QPK
also known as TLR106RL-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 960
TLR106RLS-QQSP 300, Non-Filtered300 μLMicroPointNon-filteredCase of 4800
TLR106RLS-QPK
also known as TLR106RLS-Q
QSP 300, Non-Filtered300 μLMicroPointNon-filteredPack of 960
TFLR140-200-Q200 μLMicroPointFilteredCase of 4800
TFLR140-200-QPK
also known as TFLR140-200-Q
200 μLMicroPointFilteredPack of 960
TFLR140-300-Q300 μLMicroPointFilteredCase of 4800
TFLR140-300-QPK
also known as TFLR140-300-Q
300 μLMicroPointFilteredPack of 960
TLR112NXL-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 10000
TLR112NXL-QPK
also known as TLR112NXL-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 1000
TLR112NXLR-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 4800
TLR112NXLR-QPK
also known as TLR112NXLR-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 960
TLR112NXLRS-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 4800
TLR112NXLRS-QPK
also known as TLR112NXLRS-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 960
TLR112NXLRL-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 4800
TLR112NXLRL-QPK
also known as TLR112NXLRL-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 960
TLR112NXRLS-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredCase of 4800
TLR112NXRLS-QPK
also known as TLR112NXRLS-Q
QSP 1000, Non-Filtered1000 μLExtended LengthNon-filteredPack of 960
TFLR1121000-Q1000 μLExtended LengthFilteredCase of 4800
TFLR1121000-QPK
also known as TFLR1121000-Q
1000 μLExtended LengthFilteredPack of 960
TFLR102-10-Q10 μLExtended LengthFilteredCase of 4800
TFLR102-10-QPK
also known as TFLR102-10-Q
10 μLExtended LengthFilteredPack of 960
Catalog number TLR070-QPK
also known as TLR070-Q
Price (KRW)
35,000
Pack of 1000
Description:
QSP 200, Non-Filtered
Volume (Metric):
200 μL
Tip Style:
Beveled
Filtered:
Non-filtered
Unit Size:
Pack of 1000
Price (KRW)
35,000
Pack of 1000
Dispensing certain liquids can form a film on your tip, which can affect assay accuracy and precision. Thermo Scientific™ QSP low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.
Featuring:
  • Minimize the formation of films on inside of tip
  • Reduce sample retention
  • Improve accuracy and precision
  • Improve sample recovery
  • High clarity pipette tips
  • Variety of packaging options, including environmentally friendly tip reload systems

Clean Claims:

QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.


Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
DescriptionQSP 200, Non-Filtered
Length (Metric)5.1cm
ColorNatural
FilteredNon-filtered
For Use With (Equipment)Universal Fit
FormatBulk
Product LineQSP
SterilityNon-sterile
Tip StyleBeveled
Volume (Metric)200 μL
Unit SizePack of 1000