750 Watt Ultrasonic Processors to safely process a wide range of organic and inorganic materials, from 250 microliters to 1 liter*. Typical applications include nanotechnology (producing nanoparticle materials and Graphene dispersions), cell lysing, sample preparation, homogenization, ChIP Assay, emulsification, disaggregation and deagglomeration, plus uses in the field of sonochemical processing.
- Energy and temperature set-point control
- Digital wattmeter
- Automatic tuning
- 10 hour process timer
- Independent On / Off pulser
- Variable power output control