accumet™ Liquid-Filled pH/ATC Epoxy Body Combination Electrodes - Mercury-Free
accumet™ Liquid-Filled pH/ATC Epoxy Body Combination Electrodes - Mercury-Free
accumet™ Liquid-Filled pH/ATC Epoxy Body Combination Electrodes - Mercury-Free
accumet™ Liquid-Filled pH/ATC Epoxy Body Combination Electrodes - Mercury-Free
accumet™ Liquid-Filled pH/ATC Epoxy Body Combination Electrodes - Mercury-Free
accumet™ Liquid-Filled pH/ATC Epoxy Body Combination Electrodes - Mercury-Free
Fisher Scientific™
accumet™ Liquid-Filled pH/ATC Epoxy Body Combination Electrodes - Mercury-Free
Single-junction models with Ag/AgCl reference element
Have Questions?
카탈로그 번호커넥터 유형
13620530A
ACCUMET-13620530A-EACH으로도 사용됨
BNC/Mini-phone
카탈로그 번호 13620530A
ACCUMET-13620530A-EACH으로도 사용됨
제품 가격(KRW)
-
커넥터 유형:
BNC/Mini-phone
Single-junction models with Ag/AgCl reference element
  • Rugged standard-size epoxy body
  • Single porous ceramic junction
  • Built-in ATC probe
  • BNC pH/ATC connectors
  • Rugged epoxy body for use in field or plant
  • Ideal for handheld use with aqueous samples
사양
색상Blue
충전 용액 유형SP135-500
용도(애플리케이션)Ideal for handheld use with aqueous samples
용도(장비)All Accumet XL, AR, and AB series benchtop meters
내부 참조Ag/AgCl
프로브ATC
온도(영국식 단위) 작동23°F to 212°F
온도(미터법) 범위-5°C to +80°C
커넥터 유형BNC/Mini-phone
접합부Single Ceramic
물질Epoxy
작동 온도(미터법) 작동-5°C to +80°C
유형Combination pH Electrode
pH 범위0 to 14
Unit SizeEach