사양
용량(미터법)12 mL
색상Magenta
치수(길이 x 폭)35 x 87 mm (1.38 x 3.43 in.)
용도(애플리케이션)Container for mailing, staining or storing microscope slides; Perfect for slide conveyors and specimen slide transport between the doctor's office and the lab
높이(영국식 단위)3.43 in.
높이(미터법)87 mm
물질Polypropylene
수량4 Slides
유형Slide Mailer with Envelope
Unit SizeCase of 100