Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack
Thermo Scientific™

Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack

Identify samples quickly using these PPCO amber high-profile closures. Offered in a variety of colors, these closures have no O-ring that could fall out or contaminate contents.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호색상
342825-0113Orange
342825-0110Clear
342825-0111White
342825-0112Yellow
342825-0114Green
342825-0115Red
342825-0116Blue
342825-0117Gold
342825-0118Purple
342825-0119Natural
342825-1110Teal
342825-1111Amber
카탈로그 번호 342825-0113
제품 가격(KRW)
346,000
Case of 1000
색상:
Orange
제품 가격(KRW)
346,000
Case of 1000

Quickly identify samples using Thermo Scientific™ Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials. Offered in a variety of colors, closures have no O-ring to fall out or contaminate contents.

Features:
  • Vials (listed separately) and polypropylene copolymer closures are pressure-tested together at 7.5psig.
  • Closures are for use with amber PPCO micro packaging vials to provide barrier for light-sensitive reagents.
  • Permanently attached, color-coded inserts are free of toxic heavy metals.
  • Inserts do not come in contact with vial contents.
  • Threaded, screw closure are sterile to 10-6 SAL, RNase/DNase Free, non-cytotoxic, and non-pyrogenic
  • Leakproof† when used with Nalgene Micro Packaging Vials.

Ordering Information:

Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1,000 per case. Order sterile amber vials separately (Cat. No. 342805).

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

사양
인증/적합성ISO 13485:2016
마개 재질PPCO
마개 크기11 mm
설명High-profile closures w/color coders for micro packaging vials
케이스당 수1000
팩당 수500
제품 유형Closure
마개 유형Screw Cap
색상Orange
직경(미터법) 외부11 mm
물질PPCO
제품라인Nalgene
수량500 Bag, 1000 Cs.
멸균Sterile
Unit SizeCase of 1000

자주 묻는 질문(FAQ)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.