Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Actual product may vary
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube withstands the rigors of sample preparation, transportation and short term storage, simplfying the laboratory process with a single tube.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호색상멸균최대 RCF제품 사이즈
509-GRD-A-QPK
509-GRD-A-Q으로도 사용됨
AmberNon-sterile30,000 x gPack of 500
508-GRD-G-QGreenNon-sterile30,000 x gCase of 5000
502-GRD-V-QVioletNon-sterile30,000 x gCase of 10000
502-GRD-R-QRedNon-sterile30,000 x gCase of 10000
509-GRD-V-QPK
509-GRD-V-Q으로도 사용됨
VioletNon-sterile30,000 x gPack of 500
502-GRD-V-QPK
502-GRD-V-Q으로도 사용됨
VioletNon-sterile30,000 x gPack of 1000
502-GRD-O-QPK
502-GRD-O-Q으로도 사용됨
OrangeNon-sterile30,000 x gPack of 1000
502-GRD-Y-QYellowNon-sterile30,000 x gCase of 10000
502-GRD-O-QOrangeNon-sterile30,000 x gCase of 10000
509-GRD-G-QGreenNon-sterile30,000 x gCase of 5000
509-GRD-Y-QYellowNon-sterile30,000 x gCase of 5000
509-GRD-R-QPK
509-GRD-R-Q으로도 사용됨
RedNon-sterile30,000 x gPack of 500
508-GRD-R-QPK
508-GRD-R-Q으로도 사용됨
RedNon-sterile30,000 x gPack of 500
508-GRD-V-QPK
508-GRD-V-Q으로도 사용됨
VioletNon-sterile30,000 x gPack of 500
502-GRD-B-QBlueNon-sterile30,000 x gCase of 10000
508-GRD-MIXED-QPK
508-GRD-MIXED-Q으로도 사용됨
AssortedNon-sterile30,000 x gPack of 500
509-GRD-G-QPK
509-GRD-G-Q으로도 사용됨
GreenNon-sterile30,000 x gPack of 500
502-GRDS-QNaturalSterile30,000 x gCase of 5000
509-GRD-R-QRedNon-sterile30,000 x gCase of 5000
508-GRD-R-QRedNon-sterile30,000 x gCase of 5000
509-GRD-MIXED-QPK
509-GRD-MIXED-Q으로도 사용됨
AssortedNon-sterile30,000 x gPack of 500
508-GRD-B-QBlueNon-sterile30,000 x gCase of 5000
508-GRDS-QPK
508-GRDS-Q으로도 사용됨
NaturalSterile30,000 x gPack of 250
509-GRD-A-QAmberNon-sterile30,000 x gCase of 5000
502-GRD-B-QPK
502-GRD-B-Q으로도 사용됨
BlueNon-sterile30,000 x gPack of 1000
502-GRD-A-QAmberNon-sterile30,000 x gCase of 10000
509-GRDS-QNaturalSterile30,000 x gCase of 2500
509-GRDS-QPK
509-GRDS-Q으로도 사용됨
NaturalSterile30,000 x gPack of 250
502-GRD-R-QPK
502-GRD-R-Q으로도 사용됨
RedNon-sterile30,000 x gPack of 1000
508-GRD-Y-QPK
508-GRD-Y-Q으로도 사용됨
YellowNon-sterile25,000 x gPack of 500
509-GRD-O-QOrangeNon-sterile30,000 x gCase of 5000
509-GRD-Y-QPK
509-GRD-Y-Q으로도 사용됨
YellowNon-sterile30,000 x gPack of 500
508-GRD-V-QVioletNon-sterile30,000 x gCase of 5000
502-GRD-Y-QPK
502-GRD-Y-Q으로도 사용됨
YellowNon-sterile30,000 x gPack of 1000
502-GRD-MIXED-QAssortedNon-sterile30,000 x gCase of 10000
508-GRDS-QNaturalSterile30,000 x gCase of 2500
509-GRD-B-QPK
509-GRD-B-Q으로도 사용됨
BlueNon-sterile30,000 x gPack of 500
508-GRD-G-QPK
508-GRD-G-Q으로도 사용됨
GreenNon-sterile30,000 x gPack of 500
508-GRD-Y-QYellowNon-sterile30,000 x gCase of 5000
508-GRD-O-QOrangeNon-sterile30,000 x gCase of 5000
502-GRD-MIXED-QPK
502-GRD-MIXED-Q으로도 사용됨
AssortedNon-sterile30,000 x gPack of 1000
508-GRD-MIXED-QAssortedNon-sterile30,000 x gCase of 5000
509-GRD-MIXED-QAssortedNon-sterile30,000 x gCase of 5000
502-GRD-G-QPK
502-GRD-G-Q으로도 사용됨
GreenNon-sterile30,000 x gPack of 1000
509-GRD-B-QBlueNon-sterile30,000 x gCase of 5000
508-GRD-B-QPK
508-GRD-B-Q으로도 사용됨
BlueNon-sterile30,000 x gPack of 500
502-GRD-G-QGreenNon-sterile30,000 x gCase of 10000
509-GRD-O-QPK
509-GRD-O-Q으로도 사용됨
OrangeNon-sterile30,000 x gPack of 500
508-GRD-O-QPK
508-GRD-O-Q으로도 사용됨
OrangeNon-sterile30,000 x gPack of 500
509-GRD-V-QVioletNon-sterile30,000 x gCase of 5000
카탈로그 번호 509-GRD-A-QPK
509-GRD-A-Q으로도 사용됨
제품 가격(KRW)
36,000
Pack of 500
색상:
Amber
멸균:
Non-sterile
최대 RCF:
30,000 x g
Unit Size:
Pack of 500
제품 가격(KRW)
36,000
Pack of 500
Thermo Scientific™ QSP™ Snap Cap Microcentrifuge Tube 0.6ml Graduated at 0.2, 0.4, 0.6ml, withstand up to 30,000 x g, frosted tube and cap and Amber color. Non-sterile clean claims; Rnase/Dnase, DNA, pyrogen and endotoxin free.
  • Volume range from 0.5ml to 2.0ml, available in sterile and non sterile options
  • Graduation marks
  • Colored options
  • Snap cap tube easy to open and close withstanding force up to 30,000 x G
사양
클린 클레임Non-Sterile free of: Rnase/Dnase, DNA, pyrogen and endotoxin
오토클레이브 가능Autoclavable
색상Amber
GraduatedYes
물질Polypropylene (Virgin)
최대 RCF30,000 x g
멸균Non-sterile
Unit SizePack of 500