Product Image
Thermo Scientific™
UltiMate™ 3000 Pump Piston Seals and Support Rings
Order replacement piston seals and support rings for your Thermo Scientific™ Dionex™ UltiMate™ 3000.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용도 (장비)유형
6040.9011Dionex UltiMate 3000 BX PumpPiston Seal Kit (Main Piston Seal) NP
6266.0305Dionex UltiMate 3000 RS PumpPiston Seal (Main Piston Seal) RP
6266.0304Dionex UltiMate 3000 SD PumpPiston Seal (Main Piston Seal) RP
6040.9010Dionex UltiMate 3000 BX PumpPiston Seal Kit (Main Piston Seal) RP
6025.2012Dionex UltiMate 3000 BM PumpPiston Seal Kit (Main Piston Seal)
6043.0295Dionex UltiMate 3000 XRS PumpPiston Seal Kit (Main Piston Seal) RP
카탈로그 번호 6040.9011
제품 가격(KRW)
-
용도 (장비):
Dionex UltiMate 3000 BX Pump
유형:
Piston Seal Kit (Main Piston Seal) NP
Order Thermo Scientific™ Dionex™ UltiMate™ 3000 Piston Seals and Support Rings as replacements when needed. Available individually or as kits, the seals and support rings for UltiMate 3000 pumps are specific for either normal-phase or reversed-phase chromatography. Kits include either support rings or secondary seals.
사양
용도 (장비)Dionex UltiMate 3000 BX Pump
유형Piston Seal Kit (Main Piston Seal) NP
Unit SizeEach